Ruthenium(IV) oxide, anhydrous 99.9%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ruthenium dioxide, Ruthenium tetroxide

A10816-06 A10816-03
AAA10816-06EA 427.28 USD
AAA10816-06 AAA10816-03
Ruthenium(IV) oxide, anhydrous 99.9%
Ruthenium(IV) oxide

Purity: 99.9% CAS No.: 12036-10-1 Molecular Formula: RuO2 Formula Weight: 133.07 Odor: Odorless Melting Point: 1200° subl. UN No.: 1479 MDL No.: MFCD00011210

Formula: O₂Ru
Melting Pt: 1200 °C
Density: 6.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00074857
CAS Number: 12036-10-1
EINECS: 234-840-6
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR