5-Nitro-2-furaldehyde 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Nitrofurfural
A10618-18
A10618-14
AAAA10618-18EA
232
USD
AAAA10618-18
AAAA10618-14
5-Nitro-2-furaldehyde 99%
5-Nitro-2-furaldehyde
Purity: 99%
CAS No.: 698-63-5
Molecular Formula: C5H3NO4
Formula Weight: 141.09
Flash Point:
Formula:
C₅H₃NO₄ MW: 141.08 g/mol Boiling Pt: 228 °C Melting Pt: 34…38 °C Density: 1.34 g/cm³ (20 °C) Flash Pt: 33 °C(91 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003230 CAS Number: 698-63-5 EINECS: 211-816-3 UN: 1325 ADR: "4.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...