Bismuth(III) chloride, (max. 3% H₂O) ≥98% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bismuth(III) chloride, Bismuth trichloride

A10510-14 A10510-22 A10510-36
AAAA10510-14EA 66.78 USD
AAAA10510-14 AAAA10510-22 AAAA10510-36
Bismuth(III) chloride, (max. 3% H₂O) ≥98% (dry weight)
Bismuth(III) chloride

Purity: 98% CAS No.: 7787-60-2 Molecular Formula: BiCl3 Formula Weight: 315.34 Odor: Irritating Melting Point: 230-232° Boiling Point: ca 430° subl. Merck Reference: 14,1262 UN No.: 3260 MDL No.: MFCD00003461

Formula: BiCl₃
MW: 315.34 g/mol
Boiling Pt: ∼ 430 °C (subl.)
Melting Pt: 230…232 °C
Density: 4.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003461
CAS Number: 7787-60-2
EINECS: 232-123-2
ADR: 8,III
Merck Index: 13,01261

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR