Diphenylsilanediol 98%

Supplier: Thermo Scientific Chemicals
Warning

A10492-22 A10492-36
AAAA10492-22EA 81.1 USD
AAAA10492-22 AAAA10492-36
Diphenylsilanediol 98%
Diphenylsilanediol

Purity: 98% CAS No.: 947-42-2 Molecular Formula: C12H12O2Si Formula Weight: 216.31 Odor: Odorless Melting Point: ca 150° dec. Beilstein Reference: 2523445 UN No.: 1325 MDL No.: MFCD00002101

Formula: C₁₂H₁₂O₂Si
Boiling Pt: 377 °C (1013 hPa)
Melting Pt: ∼ 150 °C (dec.)
Density: 1.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002101
CAS Number: 947-42-2
EINECS: 213-427-4
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR