Benzothiazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Benzo[d]thiazole
A10380-18
A10380-30
A10380-0B
AAA10380-18EA
30.1
USD
AAA10380-18
AAA10380-30
AAA10380-0B
Benzothiazole 97%
Benzothiazole
MDL: MFCD00005775
Beilstein: 109468
Slightly soluble in water. Soluble in alcohol, carbon disulfide
Purity: 97%
CAS No.: 95-16-9
Molecular Formula: C7H5NS
Formula Weight: 135.19
Flash Point: 107°(224°F)
Refractive Index: 1.6420
Odor: Characteristic
Melting Point: 2°
Boiling Point: 230°
Solubility: Slightly soluble in water. Soluble in alcohol, carbon disulfide
Beilstein Reference: 109468
Merck Reference: 14,1107
UN No.: 2810
MDL No.: MFCD00005775
Formula:
C₇H₅NS Boiling Pt: 230 °C Melting Pt: 2 °C Density: 1.246 g/cm³ (20 °C) Flash Pt: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005775 CAS Number: 95-16-9 EINECS: 202-396-2 UN: 2810 ADR: 6.1,III Merck Index: 13,01108 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...