Amberlite® IRN-78, Ion exchange resin cationic type (OH-) nuclear grade

Supplier: Thermo Scientific Chemicals

045591-A3 045591-36
AA45591-A3EA 168 USD
AA45591-A3 AA45591-36
Amberlite® IRN-78, Ion exchange resin cationic type (OH-) nuclear grade
Amberlite® IRN-78, Ion exchange resin cationic type (OH-)

CAS No.: 11128-95-3 Molecular Formula: Styrene-DVB; Strongly basic anion exchange resin Physical Form: Amber beads Appearance: Amber Merck Reference: 14,382 MDL No.: MFCD00145822

Storage Temperature: Ambient
MDL Number: MFCD00145822
CAS Number: 11128-95-3
Merck Index: 14,382

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR