Neodymium(III) oxide ≥99.999% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
Synonyms:
Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide
045566-22
045566-14
AA45566-22EA
769
USD
AA45566-22
AA45566-14
Neodymium(III) oxide ≥99.999% (REO, rare earth oxide basis), REacton®
Neodymium(III) oxide
Purity: 99.999%
CAS No.: 1313-97-9
Molecular Formula: Nd2O3
Formula Weight: 336.48
Physical Form: Powder
Density: 7.24
Appearance: Pale blue
Melting Point: 2272°
Merck Reference: 14,6450
MDL No.: MFCD00011134
Formula:
Nd₂O₃ MW: 336.48 g/mol Boiling Pt: 3760 °C (1013 hPa) Melting Pt: 2272 °C Density: 7.24 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011134 CAS Number: 1313-97-9 EINECS: 215-214-1 Merck Index: 14,06450 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...