Indium(III) sulphide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diindium trisulphide, Indium trisulphide

045563-18 045563-04 045563-09
AA45563-18EA 1113 USD
AA45563-18 AA45563-04 AA45563-09
Indium(III) sulphide ≥99.999% (metals basis)
Indium(III) sulphide

Purity: 99.999% CAS No.: 12030-24-9 Molecular Formula: In2S3 Formula Weight: 325.83 Physical Form: Powder Sensitivity: Moisture Sensitive MDL No.: MFCD00011061

Formula: In₂S₃
MW: 325.84 g/mol
Melting Pt: 1080
Density: 4.89 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011061
CAS Number: 12030-24-9
EINECS: 234-742-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR