Trisodium citrate, anhydrous 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium Citrate, Sodium citrate tribasic, Citric acid trisodium salt, Sodium 2-hydroxypropane-1,2,3-tricarboxylate

045556-A1 045556-30
AA45556-A1EA 125.85 USD
AA45556-A1 AA45556-30
Trisodium citrate, anhydrous 99%
Trisodium citrate

Purity: 99% CAS No.: 68-04-2 Molecular Formula: C6H5Na3O7 Formula Weight: 258.07 Physical Form: Solid Sensitivity: Hygroscopic Appearance: White Melting Point: 300° Merck Reference: 14,8602 MDL No.: MFCD00012462

Formula: C₆H₅Na₃O₇
MW: 258.07 g/mol
Melting Pt: 300 °C
Density: 1.815 g/cm³ (20 °C)
Flash Pt: 290 °C
Storage Temperature: Ambient
MDL Number: MFCD00012462
CAS Number: 68-04-2
EINECS: 200-675-3
Merck Index: 12,08746

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR