Bismuth(III) sulphide ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dibismuth trisulphide
045463-18
045463-30
AA45463-18EA
303
USD
AA45463-18
AA45463-30
Bismuth(III) sulphide ≥99.999% (metals basis)
Bismuth(III) sulphide
Purity: 99.999%
CAS No.: 1345-07-9
Molecular Formula: Bi2S3
Formula Weight: 514.15
Physical Form: Lump
Density: 7.7
Appearance: Gray
MDL No.: MFCD00014204
Formula:
Bi₂S₃ MW: 514.16 g/mol Melting Pt: 763 Density: 7.7 Storage Temperature: Ambient |
MDL Number:
MFCD00014204 CAS Number: 1345-07-9 EINECS: 215-716-0 Merck Index: 12,01329 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...