Germanium(II) telluride ≥99.999%

Supplier: Thermo Scientific Chemicals
Warning

045461-03 045461-06
AA45461-03EA 80.6 USD
AA45461-03 AA45461-06
Germanium(II) telluride ≥99.999%
Germanium(II) telluride

Purity: 99.999% CAS No.: 12025-39-7 Molecular Formula: GeTe Formula Weight: 200.19 Physical Form: -200 Mesh Powder Density: 6.14 Melting Point: 725° MDL No.: MFCD00135541

Formula: GeTe
MW: 200.21 g/mol
Melting Pt: 725
Density: 6.14
Storage Temperature: Ambient
MDL Number: MFCD00135541
CAS Number: 12025-39-7
EINECS: 234-706-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR