Indium (III) oxide ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Indium trioxide, Diindium trioxide
045460-09
045460-18
AA45460-09EA
100
USD
AA45460-09
AA45460-18
Indium (III) oxide ≥99.995% (metals basis)
Indium (III) oxide
Purity: 99.995%
CAS No.: 1312-43-2
Molecular Formula: In2O3
Formula Weight: 277.64
Physical Form: 6mm & down pieces
Density: 7.18
Melting Point: 850° volat.
Merck Reference: 14,4952
MDL No.: MFCD00011060
Formula:
In₂O₃ MW: 277.64 g/mol Melting Pt: 715.05 °C Density: 7.18 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011060 CAS Number: 1312-43-2 EINECS: 215-193-9 Merck Index: 12,04984 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...