Silver (I) sulfide ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Disilver sulphide
045458-06
045458-14
AA45458-06EA
152
USD
AA45458-06
AA45458-14
Silver (I) sulfide ≥99.999% (metals basis)
Silver(I) sulphide
Purity: 99.999%
CAS No.: 21548-73-2
Molecular Formula: Ag2S
Formula Weight: 247.80
Physical Form: 6mm & down pieces
Density: 7.317
Appearance: Dark grey to black
Odor: Odorless
Melting Point: 825°
Merck Reference: 14,8530
UN No.: 3077
MDL No.: MFCD00003406
Formula:
Ag₂S MW: 247.8 g/mol Melting Pt: 842 °C Density: 7.23 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003406 CAS Number: 21548-73-2 EINECS: 244-438-2 UN: 3077 ADR: 9,III Merck Index: 12,08677 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...