Cadmium sulfide ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cadmium (II) sulphide
045455-30
045455-18
AA45455-30EA
866
USD
AA45455-30
AA45455-18
Cadmium sulfide ≥99.999% (metals basis)
Cadmium sulphide
Purity: 99.999%
CAS No.: 1306-23-6
Molecular Formula: CdS
Formula Weight: 144.46
Physical Form: 3-12mm Pieces
Density: 4.82
Appearance: Orange
Odor: Odorless
Melting Point: 980° subl.
Merck Reference: 14,1628
UN No.: 3077
MDL No.: MFCD00010922
Formula:
CdS MW: 144.48 g/mol Melting Pt: 1400 °C Density: 4.82 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010922 CAS Number: 1306-23-6 EINECS: 215-147-8 UN: 3077 ADR: 9,III Merck Index: 12,01666 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...