Zinc sulfide ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Zinc (II) sulphide
045454-30
045454-09
045454-18
AA45454-30EA
246
USD
AA45454-30
AA45454-09
AA45454-18
Zinc sulfide ≥99.995% (metals basis)
Zinc sulphide
Purity: 99.995%
CAS No.: 1314-98-3
Molecular Formula: SZn
Formula Weight: 97.43
Physical Form: 6mm & down pieces
Density: 4.10
Appearance: White
Odor: Odorless
Melting Point: 1700°
Merck Reference: 14,10160
MDL No.: MFCD00011301
Formula:
SZn MW: 97.46 g/mol Melting Pt: 1700 °C Density: 4.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011301 CAS Number: 1314-98-3 EINECS: 215-251-3 Merck Index: 12,10294 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...