Osmium tetraoxide 2% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Osmium (VIII) oxide, Osmic acid anhydride, Osmic acid

045384-AB 045384-9W
AA45384-ABEA 141.1 USD
AA45384-AB AA45384-9W
Osmium tetraoxide 2% in aqueous solution
Osmium tetraoxide

CAS No.: 20816-12-0 Molecular Formula: OsO4 Formula Weight: 254.10 Physical Form: Liquid Appearance: Colorless to pale yellow Odor: Chlorine-like Merck Reference: 14,6893 UN No.: 3287 MDL No.: MFCD00011150

Formula: O₄Os
Storage Temperature: Ambient
MDL Number: MFCD00011150
CAS Number: 20816-12-0
EINECS: 244-058-7
UN: 3287
ADR: 6.1,III
Merck Index: 14,6893

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR