Tantalum ≥99.9% (metals basis), wire, hard, Diameter 0.203 mm (0.008 in)

Supplier: Thermo Scientific Chemicals

045133-G6 045133-G1
AA45133-G5EA 239.24 USD
AA45133-G5 AA45133-G6 AA45133-G1
Tantalum ≥99.9% (metals basis), wire, hard, Diameter 0.203 mm (0.008 in)
Tantalum

Purity: 99.9% CAS No.: 7440-25-7 Molecular Formula: Ta Formula Weight: 180.95 Physical Form: ca 0.54g/m Density: 16.69 Melting Point: 2996° Boiling Point: 5429° MDL No.: MFCD00011252

Formula: Ta
Boiling Pt: 5429 °C
Melting Pt: 2996 °C
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR