Silver 99 Metals basis, powder -120+325 mesh, atomised

Supplier: Thermo Scientific Chemicals

045097-22 045097-14
AA45097-22EA 537 USD
AA45097-22 AA45097-14
Silver 99 Metals basis, powder -120+325 mesh, atomised
Silver

Purity: 99% CAS No.: 7440-22-4 Physical Form: atomized powder, up to 10% variance on size Odor: Characteristic UN No.: 3077 MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9 III
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR