Silver ≥99.95% (metals basis), wire, hard, Diameter 0.203 mm (0.008 in)

Supplier: Thermo Scientific Chemicals

045071-G6 045071-G1 045071-G5
AA45071-G6EA 242 USD
AA45071-G6 AA45071-G1 AA45071-G5
Silver ≥99.95% (metals basis), wire, hard, Diameter 0.203 mm (0.008 in)
Silver

Purity: 99.95% CAS No.: 7440-22-4 Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR