Zinc ≥99.9% (metals basis), foil, Thickness 0.01+/-0.0025 mm (0.0004+/-0.0001 in)
Supplier: Thermo Scientific Chemicals
045059-FF
045059-FI
AA45059-FFEA
279
USD
AA45059-FF
AA45059-FI
Zinc ≥99.9% (metals basis), foil, Thickness 0.01+/-0.0025 mm (0.0004+/-0.0001 in)
Zinc
Purity: 99.9%
CAS No.: 7440-66-6
Physical Form: ≈0.045g/25x25mm
Odor: Odorless
MDL No.: MFCD00011291
Formula:
Zn Boiling Pt: 908 °C (1013 hPa) Melting Pt: 420 °C Density: 7.14 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...