Nickel chromium alloy (80:20 w%), wire, Diameter 0.2 mm (0.008 in)
Supplier: Thermo Scientific Chemicals
|
Warning
|
045057-G9
045057-H4
AA45057-H9EA
117.26
USD
AA45057-H9
AA45057-G9
AA45057-H4
Nickel chromium alloy (80:20 w%), wire, Diameter 0.2 mm (0.008 in)
Nickel chromium alloy (80:20 w%)
CAS No.: 11106-97-1
Molecular Formula: Ni:Cr; 80:20 wt%
Physical Form: ≈0.21g/m
Odor: Odorless
MDL No.: MFCD00673110
Storage Temperature:
Ambient |
MDL Number:
MFCD00673110 CAS Number: 11106-97-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...