Potassium citrate monohydrate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Potassium citrate tribasic monohydrate, Citric acid tripotassium salt monohydrate, Tripotassium citrate monohydrate, Tripotassium 2-hydroxypropane-1,2,3-tricarboxylate monohydrate
045038-36
045038-0E
AA45038-36EA
37.4
USD
AA45038-36
AA45038-0E
Potassium citrate monohydrate 97%
Potassium citrate monohydrate
Purity: 97%
CAS No.: 6100-05-6
Molecular Formula: K3C6H5O7.H2O
Formula Weight: 324.42 (306.41anhy)
Physical Form: Lump/granular
Density: 1.98
Melting Point: 230° dec.
Merck Reference: 14,7623
MDL No.: MFCD00150442
Formula:
HOC(COOK)(CH₂COOK)₂·H₂O MW: 324.41 g/mol Melting Pt: 230 °C (dec.) Density: 1.98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150442 CAS Number: 6100-05-6 EINECS: 212-755-5 Merck Index: 13,07706 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...