Palladium ≥99.9% (metals basis), wire, Diameter 0.406 mm
Supplier: Thermo Scientific Chemicals
045037-G1
045037-G5
AA45037-G1EA
516
USD
AA45037-G1
AA45037-G5
Palladium ≥99.9% (metals basis), wire, Diameter 0.406 mm
Palladium
Purity: 99.9%
CAS No.: 7440-05-3
Physical Form: ≈1.56g/m
Odor: Odorless
MDL No.: MFCD00011167
Formula:
Pd Boiling Pt: 3167 °C (1013 hPa) Melting Pt: 1555 °C Density: 12.02 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 EINECS: 231-115-6 Merck Index: 12,07121 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...