Iron(III) oxide, NanoArc®, industrial

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ferric oxide, Ferric Oxide, (iron(III) oxide), Diiron trioxide

045007-14 045007-36 045007-22
AA45007-14EA 41.91 USD
AA45007-14 AA45007-36 AA45007-22
Iron(III) oxide, NanoArc®, industrial
Iron(III) oxide

NanoArc® industrial iron oxide is a general purpose gamma-phase material suitable for use in catalytic applications such as ammonia synthesis and dehydrogenation reactions. Manufactured by growing nanocrystals rapidly in..


CAS No.: 1309-37-1 Molecular Formula: Fe2O3 Formula Weight: 159.69 Physical Form: 20-40nm APS Powder, S.A. 30-60m2/g Density: 5.24 Appearance: Red-brown Odor: Odorless Melting Point: 1565° MDL No.: MFCD00011008


NanoArc® industrial iron oxide is a general purpose gamma-phase material suitable for use in catalytic applications such as ammonia synthesis and dehydrogenation reactions. Manufactured by growing nanocrystals rapidly in..

Formula: Fe₂O₃
MW: 159.69 g/mol
Melting Pt: 1538
Density: 5.24
Storage Temperature: Ambient
MDL Number: MFCD00011008
CAS Number: 1309-37-1
EINECS: 215-168-2
Merck Index: 13,04058

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR