Cerium(IV) oxide ≥99.5% (REO, rare earth oxide basis), nanopowder

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ceric oxide, Cerium dioxide

044960-14 044960-22 044960-36
AA44960-14EA 73.04 USD
AA44960-14 AA44960-22 AA44960-36
Cerium(IV) oxide ≥99.5% (REO, rare earth oxide basis), nanopowder
Cerium(IV) oxide

Purity: 99.5% min CAS No.: 1306-38-3 Molecular Formula: CeO2 Formula Weight: 172.12 Physical Form: 15-30nm APS Powder, S.A. 30-50 m2/g Density: 7.132 Appearance: Pale yellow Melting Point: 2600° Merck Reference: 14,1989 MDL No.: MFCD00010933

Formula: CeO₂
MW: 172.11 g/mol
Boiling Pt: 3468 °C (1013 hPa)
Melting Pt: 2600 °C
Density: 7.132
Storage Temperature: Ambient
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4
Merck Index: 14,1989

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR