Zinc ≥99.9999% (metals basis), rods, Diameter 8 mm (0.31 in)

Supplier: Thermo Scientific Chemicals

044936-DJ 044936-DT
AA44936-DJEA 558 USD
AA44936-DJ AA44936-DT
Zinc ≥99.9999% (metals basis), rods, Diameter 8 mm (0.31 in)
Zinc

Purity: 99.9999% CAS No.: 7440-66-6 Odor: Odorless MDL No.: MFCD00011291

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR