Platinum iridium alloy (90:10 w%), wire, Diameter 1.5 mm (0.06 in)
Supplier: Thermo Scientific Chemicals
044812-BQ
044812-BU
AA44812-BQEA
1290
USD
AA44812-BQ
AA44812-BU
Platinum iridium alloy (90:10 w%), wire, Diameter 1.5 mm (0.06 in)
Platinum iridium alloy (90:10 w%)
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: ≈1.91g/5cm
Odor: Odorless
MDL No.: MFCD02091683
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...