Lead ≥99.995% (metals basis), wire, Diameter 0.75 mm (0.03 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
044809-G2
044809-G6
AA44809-G2EA
135
USD
AA44809-G2
AA44809-G6
Lead ≥99.995% (metals basis), wire, Diameter 0.75 mm (0.03 in)
Lead
Purity: 99.995%
CAS No.: 7439-92-1
Physical Form: ≈5g/m
Appearance: Grey
Odor: Odorless
MDL No.: MFCD00134050
Formula:
Pb Boiling Pt: 1740 °C (1013 hPa) Melting Pt: 327 °C Density: 11.3437 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134050 CAS Number: 7439-92-1 EINECS: 231-100-4 Merck Index: 12,05410 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...