Bismuth ≥99.997% (metals basis), granules 1-2 mm (0.04-0.08 in)

Supplier: Thermo Scientific Chemicals

044807-22 044807-36
AA44807-22EA 162 USD
AA44807-22 AA44807-36
Bismuth ≥99.997% (metals basis), granules 1-2 mm (0.04-0.08 in)
Bismuth

Purity: 99.997% CAS No.: 7440-69-9 Physical Form: Packaged under argon Odor: Odorless MDL No.: MFCD00134033

Formula: Bi
MW: 208.98 g/mol
Boiling Pt: 1490 °C (1013 hPa)
Melting Pt: 271 - 272 °C
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR