Urethane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Carbamic acid ethyl ester, Ethyl carbamate
044804-18
044804-30
AAA44804-18EA
43.61
USD
AAA44804-18
AAA44804-30
Urethane 98%
Urethane
Purity: 98%
CAS No.: 51-79-6
Molecular Formula: C3H7NO2
Formula Weight: 89.09
Flash Point:
Formula:
C₃H₇NO₂ MW: 89.09 g/mol Boiling Pt: 182 °C (1013 hPa) Melting Pt: 48 - 50 °C Density: 1.11 g/cm³ (20 °C) Flash Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007966 CAS Number: 51-79-6 EINECS: 200-123-1 UN: 2757 ADR: 6.1,II Merck Index: 12,10013 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...