Lead(II) sulfate, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
In pigments, galvanic batteries, lithography, weighting fabrics
CAS No.: 7446-14-2
Molecular Formula: PbSO4
Formula Weight: 303.25
Physical Form: Powder/Lump
Density: 6.2
Odor: Odorless
Merck Reference: 14,5420
UN No.: 2291
MDL No.: MFCD00011166
In pigments, galvanic batteries, lithography, weighting fabrics
Formula:
O₄PbS Melting Pt: 1170 °C Density: 6.0631 g/cm³ (18 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011166 CAS Number: 7446-14-2 EINECS: 231-198-9 UN: 2291 ADR: 6.1,III Merck Index: 12,05444 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...