Zirconium ≥99.8% (metals basis), foil, annealed, Thickness: 0.20 mm (0.008 in)

Supplier: Thermo Scientific Chemicals

044752-IB 044752-GI 044752-GR
AA44752-IBEA 898 USD
AA44752-IB AA44752-GI AA44752-GR
Zirconium ≥99.8% (metals basis), foil, annealed, Thickness: 0.20 mm (0.008 in)
Zirconium

Purity: 99.8% CAS No.: 7440-67-7 Physical Form: ≈20.3g/100x125mm, Temper: soft Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR