Platinum iridium alloy (80:20 w%), wire, hard, Diameter 1.0 mm (0.04 in)
Supplier: Thermo Scientific Chemicals
044732-BQ
044732-BS
044732-BW
AA44732-BQEA
600
USD
AA44732-BQ
AA44732-BS
AA44732-BW
Platinum iridium alloy (80:20 w%), wire, hard, Diameter 1.0 mm (0.04 in)
Platinum iridium alloy (80:20 w%)
Molecular Formula: Pt:Ir; 80:20 wt%
Physical Form: ≈0.85g/5cm
Odor: Odorless
MDL No.: MFCD02091683
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...