Cerium(IV) oxide ≥99.5% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ceric oxide, Cerium dioxide

044662-14 044662-36 044662-22
AAA44662-14EA 72.6 USD
AAA44662-14 AAA44662-36 AAA44662-22
Cerium(IV) oxide ≥99.5% (REO, rare earth oxide basis)
Cerium(IV) oxide

Purity: 99.5% CAS No.: 1306-38-3 Molecular Formula: CeO2 Formula Weight: 172.12 Physical Form: 0.07-0.1 Micron APS Powder, S. A. 8-12m2/g Density: 7.132 Appearance: Pale yellow Odor: Odorless Melting Point: 2600° Merck Reference: 14,1989 MDL No.: MFCD00010933

Formula: CeO₂
MW: 172.11 g/mol
Boiling Pt: 3468 °C (1013 hPa)
Melting Pt: 2600 °C
Density: 7.13 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4
Merck Index: 12,02035

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR