Hafnium ≥99.9% (metals basis excluding Zr), rods (Zr ∼ 3%), Diameter 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

044489-DJ 044489-DT
AA44489-DJEA 297 USD
AA44489-DJ AA44489-DT
Hafnium ≥99.9% (metals basis excluding Zr), rods (Zr ∼ 3%), Diameter 6.35 mm (0.25 in)
Hafnium

Purity: 99.9% CAS No.: 7440-58-6 Odor: Odorless MDL No.: MFCD00011032

Formula: Hf
MW: 178.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR