Titanium ≥99.2% (metals basis), foil, Thickness 1.0 mm (0.040 in)

Supplier: Thermo Scientific Chemicals
Warning

044484-GH 044484-GW
AA44484-GHEA 69.3 USD
AA44484-GH AA44484-GW
Titanium ≥99.2% (metals basis), foil, Thickness 1.0 mm (0.040 in)
Titanium

Purity: 99.2% CAS No.: 7440-32-6 Physical Form: ≈45g/100x100mm Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR