Sodium hexafluoroaluminate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trisodium hexafluoroaluminate, Sodium hexafluoroaluminate(III)
044478-14
044478-22
044478-36
AAA44478-14EA
72.6
USD
AAA44478-14
AAA44478-22
AAA44478-36
Sodium hexafluoroaluminate ≥99.9% (metals basis)
Sodium hexafluoroaluminate
Purity: 99.9%
CAS No.: 13775-53-6
Molecular Formula: Na3AlF6
Formula Weight: 209.95
Physical Form: Powder
Density: 2.90
Appearance: White
Odor: Odorless
Melting Point: 1000°
UN No.: 3260
MDL No.: MFCD00003507
Formula:
AlF₆Na₃ MW: 209.94 g/mol Melting Pt: 1000 °C Density: 2.95 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003507 CAS Number: 13775-53-6 EINECS: 237-410-6 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...