Tungsten ≥99.97% (metals basis), slug, Length: 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
044251-22
044251-14
AA44251-22EA
210
USD
AA44251-22
AA44251-14
Tungsten ≥99.97% (metals basis), slug, Length: 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)
Tungsten
Purity: 99.97%
CAS No.: 7440-33-7
Physical Form: ≈3.9g/slug
MDL No.: MFCD00011461
Formula:
W MW: 183.84 g/mol Boiling Pt: 5660 Melting Pt: 3410 Density: 19 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 Merck Index: 12,09945 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...