Sodium nitrite ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diazotizing salts

044227-22 044227-14
AA44227-22EA 491.91 USD
AA44227-22 AA44227-14
Sodium nitrite ≥99.999% (metals basis)
Sodium nitrite

Purity: 99.999% CAS No.: 7632-00-0 Molecular Formula: NaNO2 Formula Weight: 69.00 Physical Form: Crystalline Density: 2.17 Appearance: White Odor: Odorless Melting Point: 271° Merck Reference: 14,8648 UN No.: 1500 MDL No.: MFCD00011118

Formula: NaNO₂
MW: 69 g/mol
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 284 °C
Density: 2.168 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011118
CAS Number: 7632-00-0
EINECS: 231-555-9
UN: 1500
ADR: 5.1,III
Merck Index: 13,08721

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR