N,N-Dimethylformamide, anhydrous ≥99.8%, ChemSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Packaged under argon in resealable ChemSeal™ bottles.
Purity: 99.8%
CAS No.: 68-12-2
Molecular Formula: HCON(CH3)2
Formula Weight: 73.09
Flash Point: 57°(135°F)
Physical Form: Liquid
Density: 0.944
Refractive Index: 1.4310
Sensitivity: Hygroscopic
Odor: Amine-like
pH: 7
Melting Point: -61°
Boiling Point: 153°
Beilstein Reference: 605365
Merck Reference: 14,3243
UN No.: 2265
MDL No.: MFCD00003284
Formula:
HCON(CH₃)₂ Boiling Pt: 153 °C Melting Pt: –61 °C Density: 0.944 Flash Pt: 57 °C (135 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3,III REACH: 01-2119475605-32 Merck Index: 14,3243 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...