Antimony(III) bromide, ultra dry ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

043732-06 043732-14 043732-22
AAA43732-06EA 101 USD
AAA43732-06 AAA43732-14 AAA43732-22
Antimony(III) bromide, ultra dry ≥99.999% (metals basis)
Antimony(III) bromide

Purity: 99.999% CAS No.: 7789-61-9 Molecular Formula: SbBr3 Formula Weight: 361.48 Physical Form: 5-10mm Lump, Ampouled under argon Density: 4.418 Refractive Index: 1.74 Sensitivity: Air Sensitive Melting Point: 96.6° Boiling Point: 280° Merck Reference: 14,706 UN No.: 3260 MDL No.: MFCD00016317

Formula: Br₃Sb
Boiling Pt: 280 °C
Melting Pt: 96.6 °C
Density: 4.418
Storage Temperature: Ambient
MDL Number: MFCD00016317
CAS Number: 7789-61-9
EINECS: 232-179-8
UN: 3260
ADR: 8,II
Merck Index: 14,706

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR