Deuterium oxide (99.95% D) (Isotopic)

Supplier: Thermo Scientific Chemicals

043700-KV 043700-SA
AA43700-KVPK 74.6 USD
AA43700-KV AAA43700-SA
Deuterium oxide (99.95% D) (Isotopic)
Deuterium oxide

Purity: 99.95+% CAS No.: 7789-20-0 Molecular Formula: D2O Formula Weight: 20.03 Physical Form: Liquid Density: 1.105 Refractive Index: 1.338 Sensitivity: Moisture Sensitive Odor: Odorless Melting Point: 3.82° Boiling Point: 101.4° Merck Reference: 14,2940 MDL No.: MFCD00044636

Formula: D₂O
Boiling Pt: 101.3 °C (1013 hPa)
Melting Pt: 3.8 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9
Merck Index: 12,02984

Order Now

Specification Test Results

Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR