Antimony(III) chloride, ultra dry ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Antimony trichloride
043449-14
043449-22
AA43449-14EA
186
USD
AA43449-14
AA43449-22
AA43449-36
Antimony(III) chloride, ultra dry ≥99.999% (metals basis)
Antimony (III) chloride
Purity: 99.999%
CAS No.: 10025-91-9
Molecular Formula: SbCl3
Formula Weight: 228.11
Physical Form: Crystals, Ampouled under argon
Density: 3.14
Melting Point: 73.4°
Boiling Point: 223.5°
Merck Reference: 14,707
UN No.: 1733
MDL No.: MFCD00011212
Formula:
SbCl₃ MW: 228.12 g/mol Boiling Pt: 223.5 °C Melting Pt: 73.4 °C Density: 3.14 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011212 CAS Number: 10025-91-9 EINECS: 233-047-2 UN: 1733 ADR: 8,II Merck Index: 13,00713 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...