Tetrabutylammonium nitrate 98%

Supplier: Thermo Scientific Chemicals
Danger

043342-14 043342-22 043342-36 043342-06
AA43342-14EA 192 USD
AA43342-14 AA43342-22 AA43342-36 AA43342-06
Tetrabutylammonium nitrate 98%
Tetrabutylammonium nitrate

Purity: 98% CAS No.: 1941-27-1 Molecular Formula: [CH3(CH2)3]4NNO3 Formula Weight: 304.48 Physical Form: Powder Sensitivity: Hygroscopic UN No.: 1479 MDL No.: MFCD00043202

Formula: C₁₆H₃₆N₂O₃
Melting Pt: 116…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00043202
CAS Number: 1941-27-1
EINECS: 217-726-0
UN: 1479
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR