Dimethylamine 40% (w/w) in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
043261-KW
043261-KV
AA43261-KWEA
199
USD
AA43261-KW
AA43261-KV
Dimethylamine 40% (w/w) in aqueous solution
Dimethylamine
CAS No.: 124-40-3
Molecular Formula: (CH3)2NH
Formula Weight: 45.08
Flash Point: 15°(60°F)
Physical Form: Liquid in Ampoule
Density: 0.890
Refractive Index: 1.3700
Odor: Fish-like
Beilstein Reference: 605257
Merck Reference: 14,3228
UN No.: 1160
MDL No.: MFCD00008288
Formula:
C₂H₇N Density: 0.890 Flash Pt: 15 °C (60 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008288 CAS Number: 124-40-3 EINECS: 204-697-4 UN: 1160 ADR: 3,II Merck Index: 14,3228 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...