Thallium(I) iodide ≥99.9995% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

043001-09 043001-18
AAA43001-09EA 67.7 USD
AAA43001-09 AAA43001-18
Thallium(I) iodide ≥99.9995% (metals basis), Puratronic®
Thallium(I) iodide
Formula: ITl
Boiling Pt: 823 °C (1013 hPa)
Melting Pt: 440 °C
Density: 7.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011279
CAS Number: 7790-30-9
EINECS: 232-199-7
UN: 1707
ADR: 6.1,II
Merck Index: 13,09335

Order Now

Specification Test Results

Form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR