Tricyclohexylphosphine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
041952-06
041952-14
AAA41952-06EA
122
USD
AAA41952-06
AAA41952-14
Tricyclohexylphosphine 98%
Tricyclohexylphosphine
Formula:
C₁₈H₃₃P Boiling Pt: 132 - 134 °C (0.02 -rr) Melting Pt: 78 °C Density: 0.9 g/cm³ (20 °C) Storage Temperature: Freezer |
MDL Number:
MFCD00003853 CAS Number: 2622-14-2 EINECS: 220-069-2 |
Specification Test Results
Form | Crystalline |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...