Ethylene di(acetate) 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethane-1,2-diyl diacetate, Ethylene glycol diacetate, Ethylene Glycol Diacetate, 1,2-Diacetoxyethane
041910-22
041910-0E
041910-36
AA41910-22EA
21.66
USD
AA41910-22
AA41910-0E
AA41910-36
Ethylene di(acetate) 97%
Ethylene di(acetate)
Purity: 97%
CAS No.: 111-55-7
Molecular Formula: CH3CO2CH2CH2O2CCH3
Formula Weight: 146.14
Flash Point: 82°(181°F)
Physical Form: Liquid
Density: 1.13
Refractive Index: 1.4152
Melting Point: -41°
Boiling Point: 186-187°
Beilstein Reference: 1762308
Merck Reference: 14,3799
MDL No.: MFCD00008718
Formula:
C₆H₁₀O₄ MW: 146.14 g/mol Boiling Pt: 190 - 191 °C (1013 hPa) Melting Pt: -31 °C Density: 1.104 g/cm³ (20 °C) Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008718 CAS Number: 111-55-7 EINECS: 203-881-1 Merck Index: 13,03833 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...