Thionyl chloride 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Thionyl dichloride
041868-18
041868-36
041868-D3
AAA41868-18EA
125.46
USD
AAA41868-18
AAA41868-36
AAA41868-D3
Thionyl chloride 99+%
Thionyl chloride
MDL: MFCD00011449
Hydrolyzed by water forming SO2 and HCl
Purity: 99+%
CAS No.: 7719-09-7
Molecular Formula: SOCl2
Formula Weight: 118.97
Physical Form: Liquid
Density: 1.638
Refractive Index: 1.527
Sensitivity: Moisture Sensitive
Odor: Pungent
Melting Point: -104.5°
Boiling Point: 76°
Solubility: Hydrolyzed by water forming SO2 and HCl
Merck Reference: 14,9348
UN No.: 1836
MDL No.: MFCD00011449
Formula:
Cl₂OS Boiling Pt: 76 °C Melting Pt: –104.5 °C Density: 1.64 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011449 CAS Number: 7719-09-7 EINECS: 231-748-8 UN: 1836 ADR: 8,I Merck Index: 12,09485 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...