Silicon ≥99.999% (metals basis), granular virgin poly fines
Supplier: Thermo Scientific Chemicals
|
Warning
|
038542-22
038542-14
038542-36
AA38542-22EA
139
USD
AA38542-22
AA38542-14
AA38542-36
Silicon ≥99.999% (metals basis), granular virgin poly fines
Silicon
Purity: 99.999%
CAS No.: 7440-21-3
Physical Form: ~1/4in and down (fm chunks to powder)
Odor: Odorless
MDL No.: MFCD00085311
Formula:
Si Boiling Pt: 2355 °C (1013 hPa) Melting Pt: 1410 °C Density: 2.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085311 CAS Number: 7440-21-3 EINECS: 231-130-8 Merck Index: 12,08635 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...